Anti-Crebbp Rabbit Polyclonal Antibody
Supplier: Antibodies.com
Certificates
About this item
Rabbit polyclonal antibody to Crebbp for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Crebbp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77201-098
- Antigen Symbol:
- Crebbp
- Antigen Name:
- CBP
- Antigen Synonyms:
- CREB binding protein|RTS|RSTS|CBP_HUMAN|KAT3A / CBP|Crebbp|Cyclic AMP responsive enhancer binding protein|KAT3A|CREB-binding protein
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q6GQV9
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1-445 of mouse CREBBP (Q6GQV9)
- Sequence:
- MAENLLDGPPNPKRAKLSSPGFSANDNTDFGSLFDLENDLPDELIPNGELSLLNSGNLVPDAASKHKQLSELLRGGSGSSINPGIGNVSASSPVQQGLGGQAQGQPNSTNMASLGAMGKSPLNQGDSSTPNLPKQAASTSGPTPPASQALNPQAQKQVGLVTSSPATSQTGPGICMNANFNQTHPGLLNSNSGHSLMNQAQQGQAQVMNGSLGAAGRGRGAGMPYPAPAMQGATSSVLAETLTQVSPQMAGHAGLNTAQAGGMTKMGMTGTTSPFGQPFSQTGGQQMGATGVNPQLASKQSMVNSLPAFPTDIKNTSVTTVPNMSQLQTSVGIVPTQAIATGPTADPEKRKLIQQQLVLLLHAHKCQRREQANGEVRACSLPHCRTMKNVLNHMTHCQAGKACQVAHCASSRQIISHWKNCTRHDCPVCLPLKNASDKRNQQTIL
- Form:
- Liquid
- Molecular Weight:
- 290 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together