To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Indirect
- Format:Pre-coated
- Host:
- Primary antibody reactivity:HumanRatMouse
- Target protein:SYK
- Description:SYK (phospho Tyr348) cell based ELISA kit
- Size:2×96 tests
- Environmentally Preferable:
- Sample Type:Adherent cells and suspension cells
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:>5000 cells
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77205-568
Specifications
About this item
This SYK (phospho Tyr348) Cell Based ELISA kit allows for the detection of SYK (phospho Tyr348) and the effects that certain stimulation conditions have on SYK (phospho Tyr348) expression in different cell lines. Qualitative determination of SYK (phospho Tyr348) concentration is achieved by an indirect ELISA format. In essence, the SYK (phospho Tyr348) is captured by Anti-SYK (phospho Tyr348) Antibodies which in turn are detected by HRP-conjugated secondary antibodies. Through this binding, the HRP enzyme (conjugated to the secondary antibody) can catalyze a colorimetric reaction upon substrate addition. Due to the qualitative nature of this SYK (phospho Tyr348) Cell Based ELISA kit, multiple normalization methods are described: 1) Anti-GAPDH Antibody is included to serve as an internal positive control in normalizing the target absorbance values. 2) Following the colorimetric measurement of HRP activity via substrate addition, the Crystal Violet whole-cell staining method is used to determine cell density. After staining, the results can be analyzed by normalizing the absorbance values to cell amounts, by which the plating difference can be adjusted. 3) Anti-SYK Antibody is provided for normalization purposes. The absorbance values obtained for the non-phosphorylated target can be used to normalize the absorbance values for the phosphorylated target.