Order Entry
Puerto Rico
ContactUsLinkComponent
Human TMEM16A ELISA Kit
Human TMEM16A ELISA Kit
Catalog # 77201-112
Supplier:  Antibodies.com
CAS Number:  
undefined
Human TMEM16A ELISA Kit
Catalog # 77201-112
Supplier:  Antibodies.com
Supplier Number:  A312940-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    TMEM16A
  • Description:
    Human TMEM16A ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1 - 16 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.022 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77201-112

Specifications

About this item

Human TMEM16A ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TMEM16A in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1 to 16 ng/ml
  • Sensitivity: 0.022 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human TMEM16A ELISA kit (A312940) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TMEM16A in An antibody specific for TMEM16A has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TMEM16A present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-TMEM16A antibody, which also binds the TMEM16A present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-TMEM16A antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-TMEM16A antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of TMEM16A captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of TMEM16A in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.