- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Thromboxane A2 Receptor/TBXA2R
- Description:Human Thromboxane A2 receptor/TBXA2R ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:100 - 1600 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:7.45 ng/L
- Regulatory Status:RUO
- Cat. No.:77204-548
Human Thromboxane A2 Receptor/TBXA2R ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Thromboxane A2 Receptor/TBXA2R in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 100 to 1600 ng/L
- Sensitivity: 7.45 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Thromboxane A2 Receptor/TBXA2R ELISA kit (A312698) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Thromboxane A2 Receptor/TBXA2R in An antibody specific for Thromboxane A2 Receptor/TBXA2R has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Thromboxane A2 Receptor/TBXA2R present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Thromboxane A2 Receptor/TBXA2R antibody, which also binds the Thromboxane A2 Receptor/TBXA2R present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Thromboxane A2 Receptor/TBXA2R antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Thromboxane A2 Receptor/TBXA2R antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Thromboxane A2 Receptor/TBXA2R captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Thromboxane A2 Receptor/TBXA2R in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.