- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:SLC31A1/CTR1
- Description:Human SLC31A1/CTR1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:150 - 2400 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:10.41 ng/L
- Regulatory Status:RUO
- Cat. No.:77214-036
Human SLC31A1/CTR1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SLC31A1/CTR1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 150 to 2400 ng/L
- Sensitivity: 10.41 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human SLC31A1/CTR1 ELISA kit (A312414) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SLC31A1/CTR1 in serum, plasma or other biological fluids. An antibody specific for SLC31A1/CTR1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SLC31A1/CTR1 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-SLC31A1/CTR1 antibody, which also binds the SLC31A1/CTR1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SLC31A1/CTR1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SLC31A1/CTR1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of SLC31A1/CTR1 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of SLC31A1/CTR1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.