Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse SIRP alpha ELISA Kit
Mouse SIRP alpha ELISA Kit
Catalog # 77175-320
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse SIRP alpha ELISA Kit
Catalog # 77175-320
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A314199-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SIRP alpha
  • Description:
    Mouse SIRP alpha ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    150 - 2400 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    9.84 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77175-320

Specifications

About this item

Mouse SIRP alpha ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SIRP alpha in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9.84 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse SIRP alpha ELISA kit (A314199) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SIRP alpha in serum, plasma or other biological fluids. An antibody specific for SIRP alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SIRP alpha present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-SIRP alpha antibody, which also binds the SIRP alpha present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SIRP alpha antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SIRP alpha antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of SIRP alpha captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of SIRP alpha in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.