Order Entry
Puerto Rico
ContactUsLinkComponent
Human Semaphorin 3c ELISA Kit
Human Semaphorin 3c ELISA Kit
Catalog # 77173-742
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Semaphorin 3c ELISA Kit
Catalog # 77173-742
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313407-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Semaphorin 3c
  • Description:
    Human Semaphorin 3c ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    125 - 2000 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    10.23 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77173-742

Specifications

About this item

Human Semaphorin 3c ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Semaphorin 3c in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 10.23 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Semaphorin 3c ELISA kit (A313407) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Semaphorin 3c in serum, plasma or other biological fluids. An antibody specific for Semaphorin 3c has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Semaphorin 3c present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Semaphorin 3c antibody, which also binds the Semaphorin 3c present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Semaphorin 3c antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Semaphorin 3c antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Semaphorin 3c captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Semaphorin 3c in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.