- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Resistin
- Description:Mouse Resistin ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:3 - 48 ng/ml
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:0.13 ng/ml
- Regulatory Status:RUO
- Cat. No.:77173-664
Mouse Resistin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Resistin in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 3 to 48 ng/ml
- Sensitivity: 0.13 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Resistin ELISA kit (A313367) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Resistin in serum, plasma or other biological fluids. An antibody specific for Resistin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Resistin present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Resistin antibody, which also binds the Resistin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Resistin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Resistin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Resistin captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Resistin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.