- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:P-Selectin
- Description:Human P-Selectin ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:4 - 64 ng/mL
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:0.225 ng/ml
- Regulatory Status:RUO
- Cat. No.:77208-726
Human P-Selectin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human P-Selectin in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 4 to 64 ng/ml
- Sensitivity: 0.225 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human P-Selectin ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human P-Selectin in serum, plasma or other biological fluids. An antibody specific for P-Selectin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the P-Selectin present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-P-Selectin antibody, which also binds the P-Selectin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-P-Selectin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-P-Selectin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of P-Selectin captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of P-Selectin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.