Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Pepsinogen II/PGC ELISA Kit
Catalog # 77208-138
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Pepsinogen II/PGC ELISA Kit
Catalog # 77208-138
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A312170-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Pepsinogen II/PGC
  • Description:
    Mouse Pepsinogen II/PGC ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    15 - 240 ng/mL
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.58 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77208-138

Specifications

About this item

Mouse Pepsinogen II/PGC ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Pepsinogen II/PGC in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 15 to 240 ng/mL
  • Sensitivity: 0.58 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Pepsinogen II/PGC ELISA kit (A312170) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Pepsinogen II/PGC in serum, plasma or other biological fluids. An antibody specific for Pepsinogen II/PGC has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pepsinogen II/PGC present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Pepsinogen II/PGC antibody, which also binds the Pepsinogen II/PGC present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Pepsinogen II/PGC antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Pepsinogen II/PGC antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Pepsinogen II/PGC captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Pepsinogen II/PGC in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.