- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Neurotrophin 3
- Description:Human Neurotrophin 3 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:300 - 4800 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:15.12 ng/L
- Regulatory Status:RUO
- Cat. No.:77212-000
Human Neurotrophin 3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Neurotrophin 3 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 300 to 4800 ng/L
- Sensitivity: 15.12 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Neurotrophin 3 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Neurotrophin 3 in serum, plasma or other biological fluids. An antibody specific for Neurotrophin 3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Neurotrophin 3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Neurotrophin 3 antibody, which also binds the Neurotrophin 3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Neurotrophin 3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Neurotrophin 3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Neurotrophin 3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Neurotrophin 3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.