Order Entry
Puerto Rico
ContactUsLinkComponent
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Catalog # 77202-264
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Methionine Aminopeptidase 2/p67 ELISA Kit
Catalog # 77202-264
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A311431-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Methionine Aminopeptidase 2/p67
  • Description:
    Human Methionine Aminopeptidase 2/p67 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    100 - 1600 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    7.47 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77202-264

Specifications

About this item

Human Methionine Aminopeptidase 2/p67 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Methionine Aminopeptidase 2/p67 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7.47 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Methionine Aminopeptidase 2/p67 ELISA kit (A311431) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Methionine Aminopeptidase 2/p67 in serum, plasma or other biological fluids. An antibody specific for Methionine Aminopeptidase 2/p67 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Methionine Aminopeptidase 2/p67 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Methionine Aminopeptidase 2/p67 antibody, which also binds the Methionine Aminopeptidase 2/p67 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Methionine Aminopeptidase 2/p67 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Methionine Aminopeptidase 2/p67 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Methionine Aminopeptidase 2/p67 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Methionine Aminopeptidase 2/p67 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.