Human G-CSF ELISA Kit
Catalog # 77214-456
Supplier: Avantor
CAS Number:
Specifications
- Sample Type:Plasma, serum and lysates
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:16 - 2000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77214-456
Specifications
About this item
This Human G-CSF ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human G-CSF in plasma, serum, and lysates.