Troemner Flex Arm
Flex arm only, 457 mm (18) arm length
Catalog # 89083-224
Supplier: Ace Glass
Supplier #: 11058-18
Jump to:
Product Details & Documents
Nickel-plated flex arm in 457 mm (18") length.
- Supplied with flex arm only
Specifications
- Description:Flex arm only, 457 mm (18) arm length
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



