Order Entry
Puerto Rico
ContactUsLinkComponent
Troemner 3-Prong Clamp Head
Troemner 3-Prong Clamp Head
Catalog # 89083-228
Supplier:  Ace Glass
Small Business Enterprise
undefined
Troemner 3-Prong Clamp Head
Catalog # 89083-228
Supplier:  Ace Glass
Supplier Number:  11058-03
Small Business Enterprise

Specifications

  • Description:
    3-Prong head only, 69 mm (2.72") max. grip size
  • Cat. No.:
    89083-228
  • Supplier no.:
    11058-03

Specifications

About this item

Three-prong clamp heads are supplied with non-slip vinyl sleeves, and for temperatures above 100 °C, fiberglass covers.

  • Supplied with non-slip vinyl sleeves