Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium-3-mercapto-1-propanesulfonate 85%
Corrosive
Irritant
Environment
Sodium-3-mercapto-1-propanesulfonate 85%
Catalog # 77281-078
Supplier:  Ambeed
CAS Number:  
Corrosive
Irritant
Environment
Sodium-3-mercapto-1-propanesulfonate 85%
Catalog # 77281-078
Supplier:  Ambeed
Supplier Number:  A739189-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    25 g
  • Cat. No.:
    77281-078

Specifications

About this item

 :
 :178.20 g/mol
UN:3077
ADR:9,Ⅲ
 :GHS05
 :GHS07
 :GHS09
CAS Number:17636-10-1
Formula:C₃H₈O₃S₂
MDL Number:MFCD00043148