- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Metabotropic Glutamate Receptor 5
- Description:Mouse netabotropic glutamate receptor 5 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:150 - 2400 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:9.62 ng/L
- Regulatory Status:RUO
- Cat. No.:77174-898
Mouse Metabotropic Glutamate Receptor 5 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Metabotropic Glutamate Receptor 5 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 150 - 2400 ng/L
- Sensitivity: 9.62 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse Metabotropic Glutamate Receptor 5 ELISA kit (A313988) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Metabotropic Glutamate Receptor 5 in serum, plasma or other biological fluids. An antibody specific for Metabotropic Glutamate Receptor 5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Metabotropic Glutamate Receptor 5 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Metabotropic Glutamate Receptor 5 antibody, which also binds the Metabotropic Glutamate Receptor 5 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Metabotropic Glutamate Receptor 5 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Metabotropic Glutamate Receptor 5 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Metabotropic Glutamate Receptor 5 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Metabotropic Glutamate Receptor 5 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.