Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse MC1-R ELISA Kit
Mouse MC1-R ELISA Kit
Catalog # 77215-080
Supplier:  Antibodies.com
CAS Number:  
undefined
Mouse MC1-R ELISA Kit
Catalog # 77215-080
Supplier:  Antibodies.com
Supplier Number:  A312282-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MC1-R
  • Description:
    Mouse MC1-R ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    150 - 2400 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    9.63 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77215-080

Specifications

About this item

Mouse MC1-R ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse MC1-R in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 - 2400 ng/L
  • Sensitivity: 9.63 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse MC1-R ELISA kit (A312282) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse MC1-R in serum, plasma or other biological fluids. An antibody specific for MC1-R has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the MC1-R present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-MC1-R antibody, which also binds the MC1-R present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-MC1-R antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-MC1-R antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of MC1-R captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of MC1-R in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.