Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Mouse IL-1 Beta ELISA Kit

Mouse IL-1 beta ELISA kit, 96 tests
Catalog # 77174-880
Supplier:   Antibodies.com
 :  A313979-96
Mouse IL-1 Beta ELISA Kit
Catalog # 77174-880
Supplier:  Antibodies.com
CAS Number:  
Mouse IL-1 Beta ELISA Kit
Mouse IL-1 Beta ELISA Kit

Mouse IL-1 beta ELISA kit, 96 tests

CAS Number:  
Price
$591.17
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mouse IL-1 beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-1 beta in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 - 2400 ng/L
  • Sensitivity: 9.12 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse IL-1 beta ELISA kit (A313979) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IL-1 beta in serum, plasma or other biological fluids. An antibody specific for IL-1 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-1 beta present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-IL-1 beta antibody, which also binds the IL-1 beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-1 beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-1 beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of IL-1 beta captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of IL-1 beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Mouse IL-1 beta ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    150 - 2400 ng/L
  • Sample Volume:
    40 μl
  • Time to Results:
    1 h 30 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    77174-880
  • Regulatory Status:
    RUO
  • Assay duration:
    One step
  • Sensitivity:
    9.12 ng/L
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    IL-1 beta
  • Sample Type:
    Serum, plasma or other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Storage Temperature:
    4 °C

Specifications