- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:HuR/ELAVL1
- Description:Mouse HuR/ELAVL1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:100 - 1600 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:7.75 ng/L
- Regulatory Status:RUO
- Cat. No.:77209-088
Mouse HuR/ELAVL1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse HuR/ELAVL1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 100 to 1600 ng/L
- Sensitivity: 7.75 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse HuR/ELAVL1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse HuR/ELAVL1 in serum, plasma or other biological fluids. An antibody specific for HuR/ELAVL1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HuR/ELAVL1 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-HuR/ELAVL1 antibody, which also binds the HuR/ELAVL1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-HuR/ELAVL1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-HuR/ELAVL1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of HuR/ELAVL1 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of HuR/ELAVL1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.