- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Growth Hormone
- Description:Mouse growth hormone ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:20 - 320 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:0.92 ng/L
- Regulatory Status:RUO
- Cat. No.:77173-314
Mouse Growth Hormone ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Growth Hormone in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 20 to 320 ng/L
- Sensitivity: 0.92 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Mouse Growth Hormone ELISA kit (A313191) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Growth Hormone in serum, plasma or other biological fluids. An antibody specific for Growth Hormone has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Growth Hormone present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Growth Hormone antibody, which also binds the Growth Hormone present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Growth Hormone antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Growth Hormone antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Growth Hormone captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Growth Hormone in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.