- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Estrogen Receptor beta
- Description:Mouse estrogen receptor beta ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:3 - 48 ng/ml
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:0.12 ng/ml
- Regulatory Status:RUO
- Cat. No.:77210-174
Mouse Estrogen Receptor beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Estrogen Receptor beta in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 3 - 48 ng/ml
- Sensitivity: 0.12 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse Estrogen Receptor beta ELISA kit (A312145) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Estrogen Receptor beta in serum, plasma or other biological fluids. An antibody specific for Estrogen Receptor beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Estrogen Receptor beta present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Estrogen Receptor beta antibody, which also binds the Estrogen Receptor beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Estrogen Receptor beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Estrogen Receptor beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Estrogen Receptor beta captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Estrogen Receptor beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.