Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse DDIT3 ELISA Kit
Mouse DDIT3 ELISA Kit
Catalog # 77174-346
Supplier:  Antibodies.com
CAS Number:  
undefined
Mouse DDIT3 ELISA Kit
Catalog # 77174-346
Supplier:  Antibodies.com
Supplier Number:  A313710-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    DDIT3
  • Description:
    Mouse DDIT3 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    2 - 32 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.09 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77174-346

Specifications

About this item

Mouse DDIT3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse DDIT3 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 2 to 32 ng/ml
  • Sensitivity: 0.09 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse DDIT3 ELISA kit (A313710) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse DDIT3 in serum, plasma or other biological fluids. An antibody specific for DDIT3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the DDIT3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-DDIT3 antibody, which also binds the DDIT3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-DDIT3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-DDIT3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of DDIT3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of DDIT3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.