Order Entry
Puerto Rico
ContactUsLinkComponent
Human Complement Factor H-related Protein 3 ELISA Kit
Human Complement Factor H-related Protein 3 ELISA Kit
Catalog # 77207-132
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Complement Factor H-related Protein 3 ELISA Kit
Catalog # 77207-132
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A310574-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Complement Factor H-related Protein 3
  • Description:
    Human complement factor H-related protein 3 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    150 - 2400 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    9.92 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77207-132

Specifications

About this item

Human Complement Factor H-related Protein 3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement Factor H-related Protein 3 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 - 2400 ng/mL
  • Sensitivity: 9.92 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Complement Factor H-related Protein 3 ELISA kit (A310574) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement Factor H-related Protein 3 in serum, plasma or other biological fluids. An antibody specific for Complement Factor H-related Protein 3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement Factor H-related Protein 3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Complement Factor H-related Protein 3 antibody, which also binds the Complement Factor H-related Protein 3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Complement Factor H-related Protein 3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Complement Factor H-related Protein 3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Complement Factor H-related Protein 3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Complement Factor H-related Protein 3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.