Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse CaMKII Beta ELISA Kit
Mouse CaMKII Beta ELISA Kit
Catalog # 77204-902
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse CaMKII Beta ELISA Kit
Catalog # 77204-902
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A312298-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    CaMKII beta
  • Description:
    Mouse CaMKII beta ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    150 - 2400 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    9.96 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77204-902

Specifications

About this item

Mouse CaMKII beta ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CaMKII beta in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse CaMKII beta ELISA kit (A312298) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse CaMKII beta in serum, plasma or other biological fluids. An antibody specific for CaMKII beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CaMKII beta present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-CaMKII beta antibody, which also binds the CaMKII beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CaMKII beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CaMKII beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of CaMKII beta captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of CaMKII beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.