Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse AMPD2 ELISA Kit
Catalog # 77215-134
Supplier:  Antibodies.com
CAS Number:  
undefined
Mouse AMPD2 ELISA Kit
Catalog # 77215-134
Supplier:  Antibodies.com
Supplier Number:  A312259-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    AMPD2
  • Description:
    Mouse AMPD2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    125 - 2000 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    8.99 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77215-134

Specifications

About this item

Mouse AMPD2 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse AMPD2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse AMPD2 ELISA kit (A312259) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse AMPD2 in serum, plasma or other biological fluids. An antibody specific for AMPD2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the AMPD2 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-AMPD2 antibody, which also binds the AMPD2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-AMPD2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-AMPD2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of AMPD2 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of AMPD2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.