- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Adipose Triglyceride Lipase
- Description:Mouse adipose triglyceride lipase ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:50 - 800 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:3.75 ng/L
- Regulatory Status:RUO
- Cat. No.:77213-466
Mouse Adipose Triglyceride Lipase ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Adipose Triglyceride Lipase in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Adipose Triglyceride Lipase ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Adipose Triglyceride Lipase in serum, plasma or other biological fluids. An antibody specific for Adipose Triglyceride Lipase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Adipose Triglyceride Lipase present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Adipose Triglyceride Lipase antibody, which also binds the Adipose Triglyceride Lipase present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Adipose Triglyceride Lipase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Adipose Triglyceride Lipase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Adipose Triglyceride Lipase captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Adipose Triglyceride Lipase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.