- Assay duration:One step
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Acetyl Coenzyme A Carboxylase alpha
- Description:Mouse acetyl coenzyme A carboxylase alpha ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma or other biological fluids
- Detection Method:Colorimetric
- Time to Results:1 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:40 - 640 ng/L
- Storage Temperature:4 °C
- Sample Volume:40 μl
- Sensitivity:2.69 ng/L
- Regulatory Status:RUO
- Cat. No.:77208-042
Mouse Acetyl Coenzyme A Carboxylase alpha ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Acetyl Coenzyme A Carboxylase alpha in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Acetyl Coenzyme A Carboxylase alpha ELISA kit (A311085) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Acetyl Coenzyme A Carboxylase alpha in serum, plasma or other biological fluids. An antibody specific for Acetyl Coenzyme A Carboxylase alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Acetyl Coenzyme A Carboxylase alpha present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Acetyl Coenzyme A Carboxylase alpha antibody, which also binds the Acetyl Coenzyme A Carboxylase alpha present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Acetyl Coenzyme A Carboxylase alpha antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Acetyl Coenzyme A Carboxylase alpha antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Acetyl Coenzyme A Carboxylase alpha captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Acetyl Coenzyme A Carboxylase alpha in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.