Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human 5HT3B Receptor ELISA Kit
Catalog # 77209-304
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human 5HT3B Receptor ELISA Kit
Catalog # 77209-304
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A312704-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    5HT3B Receptor
  • Description:
    Human 5HT3B receptor ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    100 - 1600 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    7.59 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77209-304

Specifications

About this item

Human 5HT3B receptor ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human 5HT3B receptor in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human 5HT3B receptor ELISA kit (A312704) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human 5HT3B receptor in serum, plasma or other biological fluids. An antibody specific for 5HT3B receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the 5HT3B receptor present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-5HT3B receptor antibody, which also binds the 5HT3B receptor present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-5HT3B receptor antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-5HT3B receptor antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of 5HT3B receptor captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of 5HT3B receptor in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.