EZ-Vac 96 Vacuum Manifold
Catalog # 77020-302
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:EZ-Vac 96 vacuum manifold
- Cat. No.:77020-302
Specifications
About this item
The EZ-Vac 96 Vacuum Manifold is designed for the high-throughput, rapid parallel manual purification of nucleic acid using the ZymoPURE™ 96 Plasmid Miniprep Kit.
- Process a large number of samples simultaneously without the need for repeated centrifugation steps
The EZ-Vac 96 Vacuum Manifold is designed for the high-throughput, rapid parallel manual purification of nucleic acid using the ZymoPURE™ 96 Plasmid Miniprep Kit. The vacuum manifold allows researchers to simplify their nucleic acid purification workflows further by eliminating the need for multiple centrifugation steps through the use of a vacuum. The unique manifold design also conveniently allows the filtrate and wash solutions to be collected in an external catch flask (not included), eliminating the need for manifold disassembly and waste handling.