ZymoPURE™ Filter Plate
Supplier: Zymo Research
Certificates
About this item
The ZymoPURE™ Filter Plate can be used with centrifuges and vacuum manifolds for the rapid clarification of bacterial lysates.
- The capacity of the well is 1.0 ml
Available as a refill for the ZymoPURE™ 96 Plasmid Miniprep Kit. The ZymoPURE™ Filter Plate can be used with centrifuges and vacuum manifolds for the rapid clarification of bacterial lysates when used in conjunction with our ZymoPURE™ Plasmid Prep buffers.
Specifications
- Method / Format:Multiwell plate
- Prep size:Mini
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid