- Method / Format:Column (silica-based)
- Prep size:Scalable
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Syringe filter X
- Size:5 pack
- Cat. No.:77020-270
The ZymoPURE™ Syringe Filter features durable polypropylene construction and novel filtration media that allows for rapid clarification of up to 60 ml of neutralized bacterial lysate using the supplied polypropylene plunger.
- Rapid clarification of up to 60 ml of neutralized bacterial lysate
The ZymoPURE™ Syringe Filter features durable polypropylene construction and novel filtration media that allows for rapid clarification of up to 60 ml of neutralized bacterial lysate using the supplied polypropylene plunger. Each ZymoPURE™ Syringe Filter also includes a pre-attached ABS Luer-Lock plug in order to keep the tip clean and free from leaking during processing. Syringe filters and plungers are non-sterile and coated with silicone lubricant for easier handling. Requires the purchase of ZymoPURE™ Syringe Plungers (Cat # C1037-5) separately for use.