Order Entry
Puerto Rico
ContactUsLinkComponent
Zymo-Spin VP-S Column
Zymo-Spin VP-S Column
Catalog # 77020-308
Supplier:  Zymo Research
CAS Number:  
Zymo-Spin VP-S Column
Catalog # 77020-308
Supplier:  Zymo Research
Supplier Number:  C1083-5
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Midi
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Zymo-Spin VP-S column assembly with 15 and 50 ml reservoir
  • Size:
    5 pack
  • Binding Capacity:
    1.2 mg
  • Cat. No.:
    77020-308

Specifications

About this item

The versatile Zymo-Spin V-PS with 15 and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.

Available as a refill for the ZymoPURE™ II Plasmid Midiprep and ZymoPURE™ – Express Plasmid Midiprep Kits. The versatile Zymo-Spin V-PS with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PS features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 1.2 mg of plasmid DNA in ≥150 µl eluate when used in combination with ZymoPURE™ Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.