Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Zymo-Spin VI-PX Column

Zymo-Spin VI-PX Column

Zymo-Spin VI-PX, 5 pack
Catalog # 77020-294
Supplier:   Zymo Research
Supplier #:  C1080-5
Zymo-Spin VI-PX Column
Catalog # 77020-294
Supplier:  Zymo Research
CAS Number:  
Zymo-Spin VI-PX Column

Zymo-Spin VI-PX, 5 pack

CAS Number:  
Price
$151.39
Log in for availability
Pack of 5
Minimum order quantity is 1
Product Details & Documents

The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.

  • Exclusive to this column is a Luer-Lock bottom assembly and conical tip

The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in ≥3 ml eluate when used in combination with ZymoPURE™ Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoir.

Specifications

Specifications

  • Description:
    Zymo-Spin VI-PX
  • Size:
    5 pack
  • Cat. No.:
    77020-294
  • Prep size:
    Giga
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Bacteria
  • Binding Capacity:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.