The Zymo-Seq™ miRNA library kit is an innovative small RNA sequencing library prep kit that reduces ligation bias and provides accurate small RNA profiling from cell-free RNA (cfRNA) and total RNA.
- Accurate miRNA profiling: A unique single adapter and circularization strategy reduces ligation bias and increases miRNA detection
This simple, streamlined workflow minimizes hands-on time and uses gel-free removal of adapter dimers to generate stranded small RNA libraries from plasma, biofluids, cells, tissues, and more. To prepare Zymo-Seq™ miRNA libraries, RNA is ligated to the single miRNA adapter and the ligated product is circularized. Excess adapter is blocked and removed with specialized magnetic beads that allow a completely gel-free workflow for input as low as 1 ng. The circularized miRNA-adapter product is then reverse transcribed. The resulting cDNA is amplified using indexing PCR primers which add on illumina-compatible adapter and unique dual index (UDI) sequences for cost-effective multiplexing. Everything needed to make sequencing-ready libraries, including indexing primers and SPRI beads, are included in the kit. It's RNA-Seq made simple.
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Small RNA (miRNA, siRNA, shRNA)
- Sample type:Biofluids (serum, plasma, saliva)
- Target molecule:RNA