Anti-HMGCL Rabbit Polyclonal Antibody
Catalog # 76762-160
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:3-hydroxymethyl-3-methylglutaryl-CoA lyase
- Antigen Symbol:HMGCL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3155
- Antigen Synonyms:HL|HMGCL
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human HMGCL (NP_000182.2).
- Purification:Affinity purification
- Cat. No.:76762-160
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HMGCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat