Anti-MORF4L2 Rabbit Polyclonal Antibody
76754-408
: Prosci
- Antibody Type:Primary
- Antigen Name:Mortality factor 4 like 2
- Antigen Symbol:MORF4L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:50 µl
- Form:Liquid
- Gene ID:9643
- Antigen Synonyms:MRGX|MORF4L2|KIAA0026|RP5-1055C14.2|MORFL2
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MORF4L2 (NP_001135890.1).
- Purification:Affinity purification
- Cat. No.:76754-408
- Packaging:Vial
Type: Primary
Antigen: MORF4L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat