Anti-PCDHA13 Rabbit Polyclonal Antibody
Catalog # 76760-904
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Protocadherin alpha 13
- Antigen Symbol:PCDHA13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:56136
- Antigen Synonyms:PCDH-ALPHA13|CNRS5|CNRN5|protocadherin alpha-13|CNR5|KIAA0345-like 1|PCDH-alpha-13|ortholog of mouse CNR5|CRNR5
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 300-360 of human PCDHA13 (NP_061727.1).
- Purification:Affinity purification
- Cat. No.:76760-904
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PCDHA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat