Anti-N6AMT1 Rabbit Polyclonal Antibody
76750-368
: Prosci
- Antibody Type:Primary
- Antigen Name:N-6 adenine-specific DNA methyltransferase 1
- Antigen Symbol:N6AMT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:29104
- Antigen Synonyms:MTQ2|N-6 adenine-specific DNA methyltransferase 1 (putative)|PRED28|C21orf127|HEMK2|HemK methyltransferase family member 2|methyltransferase N6AMT1|protein N(5)-glutamine methyltransferase|m.HsaHemK2P|methylarsonite methyltransferase N6AMT1|N6AMT|N6-DNA-methyltransferase
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-186 of human N6AMT1 (NP_877426.3).
- Purification:Affinity purification
- Cat. No.:76750-368
- Packaging:Vial
Type: Primary
Antigen: N6AMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat