Anti-METTL20 Rabbit Polyclonal Antibody
Catalog # 76760-182
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Electron transfer flavoprotein subunit beta lysine methyltransferase
- Antigen Symbol:METTL20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:254013
- Antigen Synonyms:METTL2electron transfer flavoprotein beta subunit lysine methyltransferase|C12orf72|methyltransferase like 2methyltransferase-like protein 2protein N-lysine methyltransferase METTL20|ETFB-KMT|ETFB lysine methyltransferase
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 105-262 of human METTL20 (NP_001129335.1).
- Purification:Affinity purification
- Cat. No.:76760-182
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: METTL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat