Anti-CD1B Rabbit Polyclonal Antibody
Catalog # 76758-150
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CD1b molecule
- Antigen Symbol:CD1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:910
- Antigen Synonyms:T-cell surface glycoprotein CD1b|CD1b|CD1B
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 18-303 of human CD1B (NP_001755.1).
- Purification:Affinity purification
- Cat. No.:76758-150
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human