Anti-PTBP1 Rabbit Polyclonal Antibody
Catalog # 76754-298
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Polypyrimidine tract binding protein 1
- Antigen Symbol:PTBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5725
- Antigen Synonyms:PTB-1|MGC8461|PTB3|PTB-T|HNRNPI|HNRNP-I|PTB4|pPTB|MGC10830|PTB|HNRPI|PTBP1|PTB2
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PTBP1 (NP_114368.1).
- Purification:Affinity purification
- Cat. No.:76754-298
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat