Anti-SIGMAR1 Rabbit Polyclonal Antibody
Catalog # 76756-232
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Sigma non-opioid intracellular receptor 1
- Antigen Symbol:SIGMAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10280
- Antigen Synonyms:SRBP|SR31747-binding protein|Sigma1-receptor|OPRS1|Aging-associated gene 8 protein|SIGMAR1|SR-BP|Sigma non-opioid intracellular receptor 1|hSigmaR1|Sigma1R|SIG-1R|Sigma 1-type opioid receptor
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant protein of human SIGMAR1.
- Purification:Affinity purification
- Cat. No.:76756-232
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SIGMAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat