Anti-NCAM2 Rabbit Polyclonal Antibody
Catalog # 76760-718
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Neural cell adhesion molecule 2
- Antigen Symbol:NCAM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4685
- Antigen Synonyms:OCAM|MGC51008|neural cell adhesion molecule 2|NCAM2|NCAM21|HGNC:7657
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 500-690 of human NCAM2 (NP_004531.2).
- Purification:Affinity purification
- Cat. No.:76760-718
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: NCAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat