Mouse Recombinant M-CSF R Protein (from HEK293 Cells)
Catalog # 77121-674
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Mouse M-CSF Protein at 2 µg/ml (100 µl/well) can bind Mouse M-CSF R, Fc Tag with a linear range of 0.3 - 12.8 ng/ml (Routinely tested).
- Gene ID:12978
- Protein Synonyms:CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
- Protein/Peptide Name:M-CSF R
- Purity:>95% as determined by SDS-PAGE. Endotoxin level is less than 0.1 EU per µg by the LAL method.
- Molecular Weight:81.8 kDa
- Sequence:Ala 20 - Ser 511
- Formulation:Lyophilized. Tris with Glycine, Arginine and NaCl, pH 7.5.
- Shipping Temperature:RT
- Tested Applications:ELISA, WB
- Cat. No.:77121-674