Human Recombinant PD-L1 Protein (from HEK293 Cells)
Catalog # 77127-128
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human PD-L1, mouse IgG1 Fc Tag at 5 µg/ml (100 µl/well) can bind Biotinylated Human PD-1, His Tag & Fc tag with a linear range of 0.05 - 0.2 µg/ml (Routinely tested).
- Gene ID:29126
- Protein Synonyms:B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
- Protein/Peptide Name:PD-L1
- Purity:>92% as determined by SDS-PAGE. Endotoxin level is less than 0.1 EU per µg by the LAL method.
- Molecular Weight:51.6 kDa
- Sequence:Phe 19 - Arg 238
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA, WB
- Cat. No.:77127-128