SARS-CoV-2 Recombinant Spike RBD (E484Q, L452R) Protein (from Mammalian Cells)
Catalog # 77129-168
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:SARS-CoV-2
- Size:0.1 mg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze thaw cycles.Store at 2...8 °C for one week .Store at −20...−80 °C for twelve months from the date of receipt.
- Protein Synonyms:Spike RBD Protein|RBD Protein|Isolate from travelers from Inidan B.1.617
- Protein/Peptide Name:Spike RBD (E484Q, L452R)
- Molecular Weight:35 kDa
- Sequence:RBD with mutations of E484Q and L452R
- Formulation:Lyophilized. Supplied as lyophilized from PBS, pH 7.5. Please reconstitute the protein with ddH2O to the concentration with 1 mg/ml.
- Shipping Temperature:RT
- Cat. No.:77129-168