Anti-ARL6IP4 Rabbit Polyclonal Antibody
Catalog # 76748-698
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:ADP ribosylation factor like GTPase 6 interacting protein 4
- Antigen Symbol:ARL6IP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51329
- Antigen Synonyms:aip-4|SFRS2SR-25|splicing factor SRrp37|HSP-975|SRp25 nuclear protein|splicing factor|ADP-ribosylation-like factor 6 interacting protein 4|SRrp37|ADP-ribosylation factor-like 6 interacting protein 4|arginine/serine-rich 2splicing regulator SRrp38|ADP-ribosylation factor-like protein 6-interacting protein 4|SR-15|SRp25|ADP-ribosylation factor GTPase 6 interacting protein 4|HSVI binding protein|ARL-6-interacting protein 4
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 313-402 of human ARL6IP4 (NP_057722.3).
- Purification:Affinity purification.
- Cat. No.:76748-698
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ARL6IP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat