Anti-CALHM2 Rabbit Polyclonal Antibody
76756-136
: Prosci
- Antibody Type:Primary
- Antigen Name:calcium homeostasis modulator family member 2
- Antigen Symbol:CALHM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51063
- Antigen Synonyms:calcium homeostasis modulator 2|FAM26B|family with sequence similarity 26|calcium homeostasis modulator protein 2|member B
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human CALHM2 (NP_057000.2).
- Purification:Affinity purification.
- Cat. No.:76756-136
- Packaging:Vial
Type: Primary
Antigen: CALHM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse