Anti-STYXL1 Rabbit Polyclonal Antibody
Catalog # 76752-336
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:serine/threonine/tyrosine interacting like 1
- Antigen Symbol:STYXL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51657
- Antigen Synonyms:dual specificity phosphatase inhibitor MK-STYX|dual specificity phosphatase 24 (putative)|serine/threonine/tyrosine-interacting-like protein 1|MK-STYX|dual specificity protein phosphatase 24|map kinase phosphatase-like protein MK-STYX|inactive dual specificity protein phosphatase MK-STYX|DUSP24|MKSTYX
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human STYXL1 (NP_001304714.1).
- Purification:Affinity purification.
- Cat. No.:76752-336
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: STYXL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse